6-Ethoxy-2,3-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18129-25G 18129-5G
76937-272EA 481.65 USD
76937-272 76937-274
6-Ethoxy-2,3-difluorobenzaldehyde ≥97%
6-Ethoxy-2,3-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181644
CAS Number: 167684-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR