6-Bromo-7-fluoro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

17847-1G 17847-25G 17847-500MG 17847-5G
76937-238EA 155.96 USD
76937-238 76937-240 76937-242 76937-244
6-Bromo-7-fluoro-1H-indazole ≥97%
6-Bromo-7-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD23704110
CAS Number: 1427396-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR