Methyl 6-fluoro-5-methylpyridine-3-carboxylate ≥97% research grade

Supplier: AOBChem USA

Warning

17793-25G 17793-5G
76937-230EA 524.09 USD
76937-230 76937-232
Methyl 6-fluoro-5-methylpyridine-3-carboxylate ≥97% research grade
Methyl 6-fluoro-5-methylpyridine-3-carboxylate
Formula: C₈H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD11616889
CAS Number: 211122-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR