tert-Butyl benzyl(4-bromophenyl)carbamate ≥97%

Supplier: AOBChem USA

Warning

17452-25G 17452-5G
76937-198EA 275.84 USD
76937-198 76937-200
tert-Butyl benzyl(4-bromophenyl)carbamate ≥97%
tert-Butyl benzyl(4-bromophenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₈H₂₀BrNO₂
MW: 362.261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17011978
CAS Number: 693792-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR