N-Boc-4-bromo-2-fluoroaniline ≥97%

Supplier: AOBChem USA

Warning

Synonyms: N-Boc-4-Bromo-2-fluoro-aniline, tert-Butyl (4-bromo-2-fluorophenyl)carbamate

17402-25G 17402-5G
76937-188EA 204.76 USD
76937-188 76937-190
N-Boc-4-bromo-2-fluoroaniline ≥97%
N-Boc-4-bromo-2-fluoroaniline

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD07369884
CAS Number: 209958-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR