5-Hydroxymethyl-2-methylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

16950-1G 16950-500MG 16950-5G
76937-166EA 575.01 USD
76937-166 76937-168 76937-170
5-Hydroxymethyl-2-methylphenylboronic acid ≥95%
5-Hydroxymethyl-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MW: 165.982 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18827147
CAS Number: 631909-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR