6-Fluoro-2-methylnicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

16431-1G 16431-25G 16431-500MG 16431-5G
76937-116EA 350.1 USD
76937-116 76937-118 76937-120 76937-122
6-Fluoro-2-methylnicotinaldehyde ≥95%
6-Fluoro-2-methylnicotinaldehyde
Formula: C₇H₆FNO
Storage Temperature: Refrigerator
MDL Number: MFCD05662399
CAS Number: 884494-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR