6-Chloro-2-fluoro-3-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

16380-25G 16380-5G
76937-106EA 310.85 USD
76937-106 76937-108
6-Chloro-2-fluoro-3-methoxybenzoic acid ≥95%
6-Chloro-2-fluoro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD04115947
CAS Number: 886499-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR