6-Bromo-2-fluoro-3-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16291-1G 16291-25G 16291-500MG 16291-5G
76937-090EA 123.07 USD
76937-090 76937-092 76937-094 76937-096
6-Bromo-2-fluoro-3-(trifluoromethoxy)benzaldehyde ≥97%
6-Bromo-2-fluoro-3-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₃BrF₄O₂
MW: 287.006 g/mol
Storage Temperature: Refrigerator
CAS Number: 524674-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR