6-Chloro-2-fluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16279-25G 16279-5G
76937-078EA 524.09 USD
76937-078 76937-080
6-Chloro-2-fluoro-3-methylbenzaldehyde ≥97%
6-Chloro-2-fluoro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO
Storage Temperature: Refrigerator
MDL Number: MFCD01631370
CAS Number: 286474-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR