7-Bromo-2,3-dihydrobenzofuran ≥97%

Supplier: AOBChem USA

Warning

16088-25G 16088-5G
76937-070EA 445.58 USD
76937-070 76937-072
7-Bromo-2,3-dihydrobenzofuran ≥97%
7-Bromo-2,3-dihydrobenzofuran

Precaución: for Research Use Only

Formula: C₈H₇BrO
MW: 199.045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09266210
CAS Number: 206347-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR