6-Fluoro-2-methoxy-3-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15895-1G 15895-25G 15895-500MG 15895-5G
76937-050EA 207.94 USD
76937-050 76937-052 76937-054 76937-056
6-Fluoro-2-methoxy-3-methylphenylboronic acid ≥97%
6-Fluoro-2-methoxy-3-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
MW: 183.973 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526368
CAS Number: 1451391-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR