6-Bromo-2-fluoro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15606-1G 15606-25G 15606-5G
76937-016EA 62.6 USD
76937-016 76937-018 76937-020
6-Bromo-2-fluoro-3-methoxybenzaldehyde ≥97%
6-Bromo-2-fluoro-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261251
CAS Number: 853792-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR