6-Bromo-3-chloro-2-fluorobenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-3-chloro-2-fluorophenylboronic acid

15604-1G 15604-25G 15604-5G
76937-004EA 76.39 USD
76937-004 76937-006 76937-008
6-Bromo-3-chloro-2-fluorobenzeneboronic acid ≥97%
6-Bromo-3-chloro-2-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BBrClFO₂
MW: 253.261 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11111906
CAS Number: 1451392-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR