6-Chloro-2-fluoro-3-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15583-1G 15583-25G 15583-500MG 15583-5G
76936-988EA 203.7 USD
76936-988 76936-990 76936-992 76936-994
6-Chloro-2-fluoro-3-formylphenylboronic acid ≥97%
6-Chloro-2-fluoro-3-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClFO₃
MW: 202.375 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16295238
CAS Number: 1451393-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR