Ethyl 2-(2-fluoropyridin-3-yl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

152382-1G 152382-500MG 152382-5G
76936-964EA 774.46 USD
76936-964 76936-966 76936-968
Ethyl 2-(2-fluoropyridin-3-yl)-2-oxoacetate ≥97%
Ethyl 2-(2-fluoropyridin-3-yl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₉H₈FNO₃
Storage Temperature: Ambient
CAS Number: 849069-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR