6-Bromo-2-fluoro-3-isopropoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15153-25G 15153-5G
76936-952EA 240.83 USD
76936-952 76936-954
6-Bromo-2-fluoro-3-isopropoxyphenylboronic acid ≥97%
6-Bromo-2-fluoro-3-isopropoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BBrFO₃
MW: 276.895 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08457649
CAS Number: 1072951-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR