6-Bromo-2-chloro-3-methoxybenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-2-chloro-3-methoxyphenylboronic acid, (6-Bromo-2-chloro-3-methoxyphenyl)boronic acid

15150-25G 15150-5G
76936-944EA 350.1 USD
76936-944 76936-946
6-Bromo-2-chloro-3-methoxybenzeneboronic acid ≥97%
6-Bromo-2-chloro-3-methoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrClO₃
Storage Temperature: Ambient
MDL Number: MFCD09800872
CAS Number: 957062-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR