6-Bromo-2-chloro-3-methylbenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-2-chloro-3-methylphenylboronic acid, (6-Bromo-2-chloro-3-methylphenyl)boronic acid

15146-25G 15146-5G
76936-938EA 300.24 USD
76936-938 76936-940
6-Bromo-2-chloro-3-methylbenzeneboronic acid ≥97%
6-Bromo-2-chloro-3-methylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrClO₂
Storage Temperature: Ambient
MDL Number: MFCD09475910
CAS Number: 957120-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR