Ethyl 2,2-difluoro-2-(3-(trifluoromethyl)phenyl)acetate ≥97%

Supplier: AOBChem USA

Warning

147871-1G 147871-25G 147871-500MG 147871-5G
76936-904EA 123.07 USD
76936-904 76936-906 76936-908 76936-910
Ethyl 2,2-difluoro-2-(3-(trifluoromethyl)phenyl)acetate ≥97%
Ethyl 2,2-difluoro-2-(3-(trifluoromethyl)phenyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₉F₅O₂
Storage Temperature: Refrigerator
CAS Number: 175543-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR