6-Bromo-8-methylimidazo[1,2-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

14401-1G 14401-25G 14401-500MG 14401-5G
76936-864EA 153.84 USD
76936-864 76936-866 76936-868 76936-870
6-Bromo-8-methylimidazo[1,2-a]pyridine ≥97%
6-Bromo-8-methylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.059 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06496197
CAS Number: 217435-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR