2-(Hydroxymethyl)pyridine-5-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 6-(Hydroxymethyl)pyridine-3-boronic acid

13598-1G 13598-500MG 13598-5G
76936-810EA 706.56 USD
76936-810 76936-812 76936-814
2-(Hydroxymethyl)pyridine-5-boronic acid ≥97% research grade
2-(Hydroxymethyl)pyridine-5-boronic acid
Formula: C₆H₈BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09027230
CAS Number: 913835-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR