6-(4-Fluorophenyl)-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-(4-Fluorophenyl)pyridine-3-boronic acid

13471-1G 13471-25G 13471-500MG 13471-5G
76936-778EA 272.66 USD
76936-778 76936-780 76936-782 76936-784
6-(4-Fluorophenyl)-3-pyridineboronic acid ≥97%
6-(4-Fluorophenyl)-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₉BFNO₂
MW: 217.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037487
CAS Number: 1072944-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR