(6-Bromo-2-fluoro-3-propoxyphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-2-fluoro-3-propoxyphenylboronic acid

13374-25G 13374-5G
76936-724EA 262.05 USD
76936-724 76936-726
(6-Bromo-2-fluoro-3-propoxyphenyl)boronic acid ≥97% research grade
(6-Bromo-2-fluoro-3-propoxyphenyl)boronic acid
Formula: C₉H₁₁BBrFO₃
MW: 276.90 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09038422
CAS Number: 1072951-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR