Methyl 2-chloro-4-(difluoromethoxy)benzoate ≥97%

Supplier: AOBChem USA

Warning

13054-1G 13054-500MG 13054-5G
76936-702EA 968.61 USD
76936-702 76936-704 76936-706
Methyl 2-chloro-4-(difluoromethoxy)benzoate ≥97%
Methyl 2-chloro-4-(difluoromethoxy)benzoate

Precaución: for Research Use Only

Formula: C₉H₇ClF₂O₃
MW: 236.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28750638
CAS Number: 97914-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR