8-(3-(Trifluoromethyl)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%

Supplier: AOBChem USA

Warning

129978-1G 129978-25G 129978-500MG 129978-5G
76936-686EA 233.4 USD
76936-686 76936-688 76936-690 76936-692
8-(3-(Trifluoromethyl)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-(3-(Trifluoromethyl)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: for Research Use Only

Formula: C₁₅H₁₇F₃O₃
MW: 302.289 g/mol
Storage Temperature: Ambient
CAS Number: 1466429-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR