8-(3-Bromophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%

Supplier: AOBChem USA

Warning

129835-1G 129835-25G 129835-500MG 129835-5G
76936-670EA 233.4 USD
76936-670 76936-672 76936-674 76936-676
8-(3-Bromophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-(3-Bromophenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₇BrO₃
MW: 313.1869 g/mol
Storage Temperature: Ambient
CAS Number: 2002032-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR