2-Chloro-6-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-2-hydroxybenzoic acid

12944-25G 12944-5G
76936-658EA 315.09 USD
76936-658 76936-660
2-Chloro-6-hydroxybenzoic acid ≥97%
2-Chloro-6-hydroxybenzoic acid
Formula: C₇H₅ClO₃
MW: 172.5700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10687031
CAS Number: 56961-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR