Furan-2-yl(4-(trifluoromethoxy)phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

128315-1G 128315-25G 128315-500MG 128315-5G
76936-642EA 263.11 USD
76936-642 76936-644 76936-646 76936-648
Furan-2-yl(4-(trifluoromethoxy)phenyl)methanol ≥95%
Furan-2-yl(4-(trifluoromethoxy)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₂H₉F₃O₃
MW: 258.193 g/mol
Storage Temperature: Ambient
CAS Number: 1477697-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR