7-Chloro-4-methyl-1H-indole ≥97%

Supplier: AOBChem USA

Warning

124404-1G 124404-25G 124404-500MG 124404-5G
76936-606EA 186.72 USD
76936-606 76936-608 76936-610 76936-612
7-Chloro-4-methyl-1H-indole ≥97%
7-Chloro-4-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈ClN
MW: 165.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09475838
CAS Number: 941294-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR