Potassium 2-(methylsulfonyl)phenyltrifluoroborate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Potassium trifluoro(2-(methylsulphonyl)phenyl)borate, Potassium (2-methylsulphonylphenyl)trifluoroborate

12438-25G 12438-5G
76936-594EA 488.02 USD
76936-594 76936-596
Potassium 2-(methylsulfonyl)phenyltrifluoroborate ≥97%
Potassium 2-(methylsulphonyl)phenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₇H₇BF₃KO₂S
MW: 262.1000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115760
CAS Number: 850623-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR