Potassium (2,6-difluorophenyl)trifluoroboranuide ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Potassium (2,6-difluorophenyl)trifluoroborate

12414-25G 12414-5G
76936-572EA 275.84 USD
76936-572 76936-574
Potassium (2,6-difluorophenyl)trifluoroboranuide ≥97%
Potassium (2,6-difluorophenyl)trifluoroboranuide

Precaución: for Research Use Only

Formula: C₆H₃BF₅·K
MW: 219.989 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05865179
CAS Number: 267006-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR