Potassium(3-chlorophenyl)trifluoroborate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Potassium (3-chlorophenyl)trifluoroborate, (3-Chlorophenyl)trifluoroboric acid potassium salt

12410-25G 12410-5G
76936-564EA 244.01 USD
76936-564 76936-566
Potassium(3-chlorophenyl)trifluoroborate ≥95%
Potassium(3-chlorophenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₆H₄BClF₃K
MW: 218.45
Storage Temperature: Ambient
MDL Number: MFCD04115754
CAS Number: 411206-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR