6-Quinolineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Quinolin-6-ylboronic acid, Quinoline-6-boronic acid

12302-25G 12302-5G
76936-528EA 418 USD
76936-528 76936-530
6-Quinolineboronic acid ≥97%
6-Quinolineboronic acid

Precaución: for Research Use Only

Formula: C₉H₈BNO₂
Storage Temperature: Ambient
MDL Number: MFCD03093100
CAS Number: 376581-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR