Phenoxathiin-4-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: Phenoxathiin-4-ylboronic acid

12263-1G 12263-25G 12263-5G
76936-508EA 62.6 USD
76936-508 76936-510 76936-512
Phenoxathiin-4-boronic acid ≥97% research grade
Phenoxathiin-4-boronic acid
Formula: C₁₂H₉BO₃S
MW: 244.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01605731
CAS Number: 100124-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR