Potassium(3-chloro-4-fluorophenyl)trifluoroborate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Potassium (3-chloro-4-fluorophenyl)trifluoroborate, (3-Chloro-4-fluorophenyl)trifluoroboric acid potassium salt

12140-25G 12140-5G
76936-492EA 187.78 USD
76936-492 76936-494
Potassium(3-chloro-4-fluorophenyl)trifluoroborate ≥95%
Potassium(3-chloro-4-fluorophenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₆H₃BClF₄K
MW: 236.44
Storage Temperature: Ambient
MDL Number: MFCD04115752
CAS Number: 850623-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR