N-(2,5-Difluoro-4-nitrophenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

120931-1G 120931-25G 120931-5G
76936-482EA 100.79 USD
76936-482 76936-484 76936-486
N-(2,5-Difluoro-4-nitrophenyl)acetamide ≥97%
N-(2,5-Difluoro-4-nitrophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₆F₂N₂O₃
MW: 216.142 g/mol
Storage Temperature: Ambient
CAS Number: 366-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR