Methyl-6-fluoronicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Fluoronicotinic acid methyl ester, Methyl 6-fluoropyridine-3-carboxylate

12008-25G 12008-5G
76936-476EA 183.54 USD
76936-476 76936-478
Methyl-6-fluoronicotinate ≥97%
Methyl-6-fluoronicotinate

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
Storage Temperature: Ambient
MDL Number: MFCD03095098
CAS Number: 1427-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR