6-Phenylpyridine-3-boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Borono-2-phenylpyridine, (6-Phenylpyridin-3-yl)boronic acid

11646-1G 11646-25G 11646-500MG 11646-5G
76936-462EA 198.39 USD
76936-462 76936-464 76936-466 76936-468
6-Phenylpyridine-3-boronic acid ≥97%
6-Phenylpyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD03788243
CAS Number: 155079-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR