6-Benzyloxy-2-naphthaleneboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 6-Benzyloxy-2-naphthylboronic acid, 6-(Benzyloxy)-2-naphthylboronic acid

11070-25G 11070-5G
76936-434EA 524.09 USD
76936-434 76936-436
6-Benzyloxy-2-naphthaleneboronic acid ≥95%
6-Benzyloxy-2-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₅BO₃
MW: 278.110 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115402
CAS Number: 152915-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR