Methyl 2-chloro-5-methoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

108612-1G 108612-25G 108612-500MG 108612-5G
76936-424EA 222.79 USD
76936-424 76936-426 76936-428 76936-430
Methyl 2-chloro-5-methoxynicotinate ≥97%
Methyl 2-chloro-5-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
Storage Temperature: Ambient
CAS Number: 1256791-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR