6-(1,3-Dioxolan-2-yl)picolinic acid ≥97%

Supplier: AOBChem USA

Warning

107969-1G 107969-25G 107969-500MG 107969-5G
76936-408EA 224.92 USD
76936-408 76936-410 76936-412 76936-414
6-(1,3-Dioxolan-2-yl)picolinic acid ≥97%
6-(1,3-Dioxolan-2-yl)picolinic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MW: 195.172 g/mol
Storage Temperature: Ambient
CAS Number: 1546229-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR