Potassium (3-acetylphenyl)trifluoroborate ≥97% research grade

Supplier: AOBChem USA

Warning

10525-25G 10525-5G
76936-396EA 215.37 USD
76936-396 76936-398
Potassium (3-acetylphenyl)trifluoroborate ≥97% research grade
Potassium (3-acetylphenyl)trifluoroborate
Formula: C₈H₇BF₃O·K
MW: 226.045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368352
CAS Number: 854906-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR