Methyl-4-bromo-2,6-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 4-bromo-2,6-dimethylbenzoate

10394-25G 10394-5G
76936-360EA 537.88 USD
76936-360 76936-362
Methyl-4-bromo-2,6-dimethylbenzoate ≥97%
Methyl-4-bromo-2,6-dimethylbenzoate
Formula: C₁₀H₁₁BrO₂
MW: 243.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11227152
CAS Number: 90841-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR