piperidin-1-yl(pyridin-4-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

102943-1G 102943-25G 102943-5G
76936-352EA 89.12 USD
76936-352 76936-354 76936-356
piperidin-1-yl(pyridin-4-yl)methanone ≥97%
piperidin-1-yl(pyridin-4-yl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O
MW: 190.242 g/mol
Storage Temperature: Ambient
CAS Number: 4968-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR