5-Chloro-2,3-dimethoxy-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

95112-1G 95112-25G 95112-500MG 95112-5G
76936-124EA 234.46 USD
76936-124 76936-126 76936-128 76936-130
5-Chloro-2,3-dimethoxy-4-(methylthio)pyridine ≥97%
5-Chloro-2,3-dimethoxy-4-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO₂S
MW: 219.689 g/mol
Storage Temperature: Refrigerator
CAS Number: 2918958-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR