5-Fluoro-8-methylquinoline ≥97%

Supplier: AOBChem USA

Warning

85782-1G 85782-25G 85782-500MG 85782-5G
76936-060EA 123.07 USD
76936-060 76936-062 76936-064 76936-066
5-Fluoro-8-methylquinoline ≥97%
5-Fluoro-8-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈FN
Storage Temperature: Ambient
CAS Number: 88474-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR