5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(p-tolyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

85431-1G 85431-25G 85431-500MG 85431-5G
76936-044EA 260.99 USD
76936-044 76936-046 76936-048 76936-050
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(p-tolyl)pyridine ≥97%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(p-tolyl)pyridine

Precaución: for Research Use Only

Formula: C₁₈H₂₂BNO₂
MW: 295.184 g/mol
Storage Temperature: Ambient
CAS Number: 2223028-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR