5-Bromo-4-chloro-2-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

80559-1G 80559-25G 80559-500MG 80559-5G
76936-004EA 233.4 USD
76936-004 76936-006 76936-008 76936-010
5-Bromo-4-chloro-2-fluorophenol ≥97%
5-Bromo-4-chloro-2-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrClFO
Storage Temperature: Ambient
CAS Number: 1807224-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR