5-Bromo-3-chloro-2-isopropoxypyridine ≥97%

Supplier: AOBChem USA

Warning

79819-1G 79819-25G 79819-500MG 79819-5G
76935-988EA 217.49 USD
76935-988 76935-990 76935-992 76935-994
5-Bromo-3-chloro-2-isopropoxypyridine ≥97%
5-Bromo-3-chloro-2-isopropoxypyridine

Precaución: for Research Use Only

Formula: C₈H₉BrClNO
Storage Temperature: Ambient
CAS Number: 1211531-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR